missing translation for 'onlineSavingsMsg'
Learn More

PLK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10977-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827723

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytokine-inducible kinase is a putative serine/threonine kinase. CNK contains both a catalytic domain and a putative regulatory domain. It may play a role in regulation of cell cycle progression and tumorigenesis.
Specifications

Specifications

PLK3
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q60806, Q9H4B4, Q9R011
CNK, FGF inducible kinase, FNK, PLK 3, PLK3, Polo like kinase 3, PRK, Proliferation related kinase
Rabbit
IgG
20 μL
-20°C
Primary
1263, 12795, 58936
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PLK3
Liquid
Plk3
PLK3 Fusion Protein Ag1431
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers