Läs mer
PLOD3 Mouse anti-Human, CoraLite 594, Clone: 2G12B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60058-100UL
Beskrivning
The PLOD3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLOD3. This antibody recognizes human antigen. The PLOD3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifikationer
| PLOD3 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence | |
| 2G12B8 | |
| CoraLite 594 | |
| 100 μL | |
| 8985 | |
| IgG2a |