missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11027-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.Specifications
PLOD3 | |
Polyclonal | |
Unconjugated | |
PLOD3 | |
LH3, Lysyl hydroxylase 3, PLOD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26433, 288583, 8985 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60568, Q5U367, Q9R0E1 | |
Plod3 | |
PLOD3 Fusion Protein Ag1480 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |