Läs mer
Tocris Bioscience™ PLpro inhibitor 6
Beskrivning
PLpro inhibitor 6 is a SARS-CoV and SAR-CoV-2 PLpro inhibitor (IC50 values are 2.6 and 5.0 μM, respectively). Inhibits SARS-CoV and SARS-CoV-2 infection of Vero E6 cells in vitro (EC50 values are 13.1 and 21 μM, respectively).
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5-(Acetylamino)-2-methyl-N-[(1 R)-1-(1-naphthalenyl)ethyl]benzamide |
| CAS | 1093070-14-4 |
| Kvantitet | 25 mg |
| Mål | Papain-like Protease Inhibitors |
| Molekylformel | C22H22N2O2 |
| Renhet | 0.98 |