Läs mer
Pluripotin, MedChemExpress
Produktkod. 16470116
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
50mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16470116 2 mg -
16465036 100 mg -
16454058 10 mg 10mg
16452936 25 mg -
16436757 5 mg -
16410417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16470116 Leverantör Medchem Express Leverantörsnummer HY10579
MedChemExpress Pluripotin is a dual inhibitor of ERK1 and RasGAP with KDs of 98 nM and 212 nM, respectively. Pluripotin also inhibits RSK1, RSK2, RSK3, and RSK4 with IC50s of 0.5, 2.5, 3.3, and 10.0 μM, respectively.
Pluripotin is a dual inhibitor of ERK1 and RasGAP with KDs of 98 nM and 212 nM, respectively. Pluripotin also inhibits RSK1, RSK2, RSK3, and RSK4 with IC50s of 0.5, 2.5, 3.3, and 10.0 μM, respectively.
Specifikationer
Kemiskt namn eller material Pluripotin
Fysisk form Solid
CAS 839707-37-8
Färg White
Kvantitet 2 mg
Anteckningar om renhetsgrad Research
Molekylformel C27H25F3N8O2
För användning med (applikation) Cancer-Kinase/protease
Synonym SC1
Löslighetsinformation DMSO : ≥ 33 mg/mL (59.94 mM)
LEDER CN1C(NC2=NC=C3C(N(C)C(N(C4=CC(NC(C5=CC=CC(C(F)(F)F)=C5)=O)=CC=C4C)C3)=O)=N2)=CC(C)=N1
Molekylvikt (g/mol) 550.54
Formel vikt 550.54
Procent renhet 98.19%