Learn More
PLXNB2 Mouse anti-Human, Unconjugated, Clone: 2G9B2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67265-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839795
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The PLXNB2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLXNB2. This antibody recognizes human, mouse, rat antigen. The PLXNB2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PLXNB2
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Immunohistochemistry, Western Blot
2G9B2
Unconjugated
150 μL
23654
IgG1