Läs mer
Invitrogen™ PMCA ATPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113788
Beskrivning
Immunogen sequence: GDMANSSIEF HPKPQQQRDV PQAGGFGCTL AELRTLMELR GAEALQKIEE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%.
The calcium pump of the plasma membrane (PMCA ATPase) pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the sarcoplasmic and endoplasmic reticulum calcium (SERCA) ATPase and the sodium/potassium pump. Studies indicate that there are four known isoforms of the PMCA ATPase which are encoded by a multigene family.
Specifikationer
| PMCA ATPase | |
| Polyclonal | |
| Unconjugated | |
| ATP2B1 | |
| 2810442I22Rik; 6430519O13Rik; ATP2B; ATP2B1; ATP2B2; ATP2B3; ATP2B4; ATPase isoform 2 Na+K+ transporting beta polypeptide 2; ATPase isoform 2, Na+K+ transporting, beta polypeptide 2; ATPase plasma membrane Ca2+ transporting 1; ATPase plasma membrane Ca2+ transporting 2; ATPase plasma membrane Ca2+ transporting 3; ATPase plasma membrane Ca2+ transporting 4; ATPase, Ca++ transporting, plasma membrane 1; ATPase, Ca++ transporting, plasma membrane 2; ATPase, Ca++ transporting, plasma membrane 3; ATPase, Ca++ transporting, plasma membrane 4; ATP-dependent plasma membrane calcium transporter; Ca2+/Mg2+ ATPase; CFAP39; cilia and flagella associated protein 39; CLA2; D6Abb2e; deaf waddler; dfw; E130111D10Rik; Gena300; jog; Matrix-remodeling-associated protein 1; MXRA1; OPCA; plasma membrane Ca(2+)-ATPase; plasma membrane Ca++-ATPase; plasma membrane Ca2+ pump 2; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium ATPase isoform 1b; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium ATPase isoform 4a; plasma membrane calcium ATPase isoform 4b; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; plasma membrane calcium pump isoform 1b; Plasma membrane calcium pump isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 1; plasma membrane calcium-transporting ATPase 2; Plasma membrane calcium-transporting ATPase 3; plasma membrane calcium-transporting ATPase 4; plasma membrane calcium-transporting ATPase 4; plasma membrane calcium-transporting ATPase 1; PMCA; PMCA1; Pmca1a; Pmca1b; Pmca1c; PMCA1kb; PMCA2; PMCA2a; PMCA2i; Pmca3; PMCA3a; Pmca4; PMCA4a; PMCA4b; PMCA4x; sarcolemmal calcium pump; SCAX1; Tmy; wms; wri; wriggle | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 490, 491, 492, 493 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P20020, P23634, Q01814, Q16720 | |
| ATP2B1, ATP2B2, ATP2B3, ATP2B4 | |
| Synthetic peptide directed towards the N terminal region of human ATP2B3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.