missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ PMCA2 ATPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593257
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: MGDMTNSDFY SKNQRNESSH GGEFGCTMEE LRSLMELRGT EAVVKIKETY GDTEAICRRL KTSPVEGLPG TAPDLEKRKQ; Positive Samples: Mouse ovary, Mouse lung, Mouse liver, Rat liver; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, synapse.
The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin.
Specifikationer
| PMCA2 ATPase | |
| Polyclonal | |
| Unconjugated | |
| ATP2B2 | |
| Atp2b2; ATPase isoform 2 Na+K+ transporting beta polypeptide 2; ATPase isoform 2, Na+K+ transporting, beta polypeptide 2; ATPase plasma membrane Ca2+ transporting 2; ATPase, Ca++ transporting, plasma membrane 2; D6Abb2e; deaf waddler; dfw; Gena300; jog; plasma membrane Ca(2+)-ATPase; plasma membrane Ca++-ATPase; plasma membrane Ca2+ pump 2; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 2; plasma membrane calcium pump; Plasma membrane calcium pump isoform 2; Plasma membrane calcium-transporting ATPase 2; PMCA2; PMCA2a; PMCA2i; Tmy; wms; wri; wriggle | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11941, 24215, 491 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.56 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11506, Q01814, Q9R0K7 | |
| ATP2B2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human ATP2B2 (NP_0016742). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering