missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10666-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the isomerization of mannose 6-phosphate to mannose 1-phosphate, which is a precursor to GDP-mannose necessary for the synthesis of dolichol-P-oligosaccharides. Mutations in this gene have been shown to cause defects in glycoprotein biosynthesis, which manifests as carbohydrate-deficient glycoprotein syndrome type I.Specifications
PMM2 | |
Polyclonal | |
Unconjugated | |
PMM2 | |
CDG1, CDG1a, CDGS, phosphomannomutase 2, PMM 2, PMM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
302915, 5373, 54128 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15305, Q9Z2M7 | |
PMM2 | |
PMM2 Fusion Protein Ag1031 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |