Learn More
BRAND™ PMP Low Form Graduated Beakers with Embossed Scale
Click to view available options
Capacity (Metric):
50 mL
100 mL
500 mL
1000 mL
5000 mL
Description
- Clear PMP (Polymethylpentene)
- Embossed graduations, beaded rim and spout
- Adheres to ISO 7056
- Autoclavable (121°C)
Specifications
Color | Clear |
Capacity (Metric) | 50 mL |
Material | PMP |
Includes | Beaded Rim with Spout |
Increments | 10 mL |
Quantity | 4 Pk. |
Autoclavable | Autoclavable |
Graduated | Yes |
Certifications/Compliance | ISO 7056 |
Scale | Embossed Scale |