missing translation for 'onlineSavingsMsg'
Learn More

PMP2 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12717-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840594

  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of domit demyelinating CMT neuropathy.
Specifications

Specifications

PMP2
Polyclonal
Unconjugated
PMP2
FABP8, M FABP, MP2, Myelin P2 protein, P2, peripheral myelin protein 2, PMP2
Rabbit
Antigen Affinity Chromatography
RUO
100151890, 18857, 5375, 688790
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.54 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P02689, P24526, P86412
Pmp2
PMP2 Fusion Protein Ag3411
150 μL
Primary
Rat, Porcine, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers