missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMPCB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PMPCB |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835544
|
Proteintech
16064-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825544
|
Proteintech
16064-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.Specifications
PMPCB | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O75439, Q03346, Q9CXT8 | |
64198, 73078, 9512 | |
PMPCB Fusion Protein Ag8937 | |
Primary | |
-20°C | |
PMPCB |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Beta MPP, MPP11, MPPB, MPPP52, P 52, PMPCB | |
Pmpcb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |