missing translation for 'onlineSavingsMsg'
Learn More

PMPCB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16064-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825544

  • 3313.85 SEK / 150µL
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
Specifications

Specifications

PMPCB
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75439, Q03346, Q9CXT8
Beta MPP, MPP11, MPPB, MPPP52, P 52, PMPCB
Rabbit
IgG
150 μL
-20°C
Primary
64198, 73078, 9512
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PMPCB
Liquid
Pmpcb
PMPCB Fusion Protein Ag8937
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers