Läs mer
Invitrogen™ PNO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110188
Beskrivning
Immunogen sequence: MESEMETQSA RAEEGFTQVT RKGGRRAKKR QAEQLSAAGE GGDAGRMDTE EARPAKRPVF PPLCGDGLLS GKEETRKIPV PANRYTPLKE NWMKIFTPIV EHLGLQIRFN LKSRNVEIRT CKETKDVSAL TKAADFVKAF ILGFQVEDAL ALIRLDDLFL ESFEITDVKP LKGDHLSRAI GRIAGKGGKT KFTIENVTRT RIVLADVKVH ILGSFQNIKM ARTAICNLIL GNPPSKVYGN IRAVASRSAD RF.
Positively regulates dimethylation of two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA (PubMed:25851604). [UniProt].
Specifikationer
| PNO1 | |
| Polyclonal | |
| Unconjugated | |
| PNO1 | |
| 1810003N24Rik; AA407421; AU015468; AU019495; AU042323; Emi3; expressed during mesenchymal induction 3; Imi3; induced during mesenchymal induction 3; KHRBP1; KH-type RNA-binding protein 1; partner of NOB1 homolog; partner of NOB1 homolog (S. cerevisiae); Pno1; putatative 28 kDa protein; putative 28 kDa protein; RNA binding protein; RNA-binding protein PNO1; RRP20 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 289809, 56902 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6VBQ8, Q9NRX1 | |
| PNO1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-252 of human PNO1 (NP_064528.1). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.