Läs mer
Podophyllotoxin 4-O-Glucoside, TRC
Produktkod. 30422196
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30422196 1 mg
1 options
Produktkod. 30422196 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP6810101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Podophyllotoxin 4-O-Glucoside
CAS 16481-54-2
Molekylformel C35 H36 O13
Kvantitet 1 mg
Synonym (5R,5aR,8aR,9R)-9-(β-D-Glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(3,4,5-trimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, Podophyllotoxin','β-D-glucopyranoside, NSC 163024, Podophyllotoxin 1-O-β-D-Glycoside, Podophyllotoxin 3-β-D-Glucopyranoside, Podophyllotoxin 4-O-Glucoside, Podophyllotoxin 7'-O-β-D-Glucopyranoside, Podophyllotoxin β-D-Glucoside, Podophyllotoxin-4-O-β-D-Glucopyranoside
InChI formel InChI=1S/C35H36O13/c1-39-23-9-17(10-24(40-2)31(23)41-3)26-18-11-21-22(45-15-44-21)12-19(18)30(20-13-42-33(38)27(20)26)47-35-29(37)28(36)32-25(46-35)14-43-34(48-32)16-7-5-4-6-8-16/h4-12,20,25-30,32,34-37H,13-15H2,1-3H3/t20-,25+,26+,27-,28+,29+,30-,32+,34?,35-/m0/s1
LEDER COc1cc(cc(OC)c1OC)[C@H]2[C@@H]3[C@H](COC3=O)[C@@H](O[C@@H]4O[C@@H]5COC(O[C@H]5[C@H](O)[C@H]4O)c6ccccc6)c7cc8OCOc8cc27
IUPAC-namn (5R,5aR,8aR,9R)-5-[[(4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(3,4,5-trimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
Molekylvikt (g/mol) 664.65
Formel vikt 664.2156