missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POFUT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14929-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
POFUT1, an endoplasmic reticulum-residing member of the glycosyltransferase O-Fuc family, adds O-fucose through an O-glycosidic linkage to conserved serine or threonines in epidermal growth factor-like repeats of several cell surface and secreted proteins. Unlike its homolog POFUT2, POFUT1 can also catalyze the transfer of fucose to thrombospondin type 1 repeats. Many of the substrates of POFUT1 are involved in ligand-induced receptor signaling. One such protein is Notch; mouse ES cells lacking POFUT have normal levels of Notch receptors at the cell surface, but these receptors do not bind Notch ligands or exhibit Notch signaling.Specifications
POFUT1 | |
Polyclonal | |
Unconjugated | |
POFUT1 | |
FUT12, KIAA0180, O Fuc T, O FucT 1, O FUT, Peptide O fucosyltransferase 1, POFUT1, protein O fucosyltransferase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140484, 23509, 311551 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6EV70, Q91ZW2, Q9H488 | |
Pofut1 | |
POFUT1 Fusion Protein Ag6710 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |