missing translation for 'onlineSavingsMsg'
Learn More

POLD4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26209-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824735

  • 1236.20 SEK / 20µL
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes the smallest subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3′ to 5′ exonuclease activity and plays a critical role in DNA replication and repair. The encoded protein enhances the activity of DNA polymerase delta and plays a role in fork repair and stabilization through interactions with the DNA helicase Bloom syndrome protein. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications

Specifications

POLD4
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9HCU8
DNA polymerase delta subunit 4, p12, POLD4, POLDS
Rabbit
IgG
20 μL
-20°C
Primary
57804
Western Blot
Unconjugated
POLD4
Liquid
POLD4
POLD4 Fusion Protein Ag23924
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers