missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLDIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15080-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
POLDIP2 is a protein that interacts with the DNA polymerase delta p50 subunit, as well as with proliferating cell nuclear antigen. The encoded protein maybe play a role in the ability of the replication fork to bypass DNA lesions. Alternative splicing results in multiple transcript variants.Specifications
POLDIP2 | |
Polyclonal | |
Unconjugated | |
POLDIP2 | |
PDIP38, POLDIP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26073, 287544, 67811 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91VA6, Q9Y2S7 | |
Poldip2 | |
POLDIP2 Fusion Protein Ag7154 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |