missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14873-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.Specifications
POLM | |
Polyclonal | |
Unconjugated | |
POLM | |
DNA polymerase mu, Pol Mu, POLM, polymerase (DNA directed), mu, Tdt N | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27434 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NP87 | |
POLM | |
POLM Fusion Protein Ag6642 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |