missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2I Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17270-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
POLR2I is a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with two other polymerase subunits, forms the DNA binding domain of the polymerase, a groove in which the DNA template is transcribed into RNA. POLR2I has two zinc finger motifs with conserved cysteines and the subunit does possess zinc binding activity. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests. This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with two other polymerase subunits, forms the DNA binding domain of the polymerase, a groove in which the DNA template is transcribed into RNA. The product of this gene has two zinc finger motifs with conserved cysteines and the subunit does possess zinc binding activity.Specifications
POLR2I | |
Polyclonal | |
Unconjugated | |
POLR2I | |
hRPB14.5, POLR2I, RNA polymerase II subunit B9, RPB14.5, RPB9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
292778, 5438, 69920 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36954, P60898 | |
POLR2I | |
POLR2I Fusion Protein Ag11027 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |