missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | POLR2L |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835114
|
Proteintech
15779-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825114
|
Proteintech
15779-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains four conserved cysteines characteristic of an atypical zinc-binding domain. Like its counterpart in yeast, this subunit may be shared by the other two DNA-directed RNA polymerases.Specifications
POLR2L | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62875, P62876 | |
502374, 5441, 66491 | |
POLR2L Fusion Protein Ag8468 | |
Primary | |
-20°C | |
POLR2L |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hRPB7.6, hsRPB10b, POLR2L, RBP10, RPABC5, RPB10, RPB10 homolog, RPB10beta, RPB7.6 | |
POLR2L | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |