Läs mer
Poly(4-hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol-alt-1,4-butanedioic Acid) (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Plastic additive 22 |
| CAS | 65447-77-0 |
| Molekylformel | (C15H25NO4)n |
| Kvantitet | 500 mg |
| Synonym | 1-Piperidineethanol','4-hydroxy-2,2,6,6-tetramethyl-','polymer with dimethyl butanedioate (9CI), Butanedioic acid','dimethyl ester','polymer with 4-hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol (9CI), 1-(2-Hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol-dimethyl succinate copolymer, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinethanol-dimethyl succinate copolymer, 622LD, BLS 1622, Butanedioic acid dimethyl ester-4-hydroxy-2,2,6,6-tetramethyl-1-piperidine ethanol copolymer, BW 10LD, Chemsorb LS 622, Chiguard 622LD, Chimassorb 622LD, Chisorb 622LD, Dimethyl succinate-1-(2-hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol copolymer, Dimethyl succinate-1-(2-hydroxyethyl)-4-hydroxy-2,2,6,6-tetramethylpiperidine copolymer, Dimethyl succinate-2-(4-hydroxy-2,2,6,6-tetramethyl-1-piperidyl)ethanol copolymer, Dimethyl succinate-4-hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine copolymer, Dimethyl succinate-4-hydroxy-2,2,6,6,-tetramethyl-1-piperidineethanol, Dimethyl succinate-4-hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol copolymer, Dimethyl succinate-4-hydroxy-2,2,6,6-tetramethyl-1-piperidylethanol copolymer, Hindered amine 622, Jylstab 788, Kemistab 62, Light stabilizer 622LD, Light Stabilizer 788, Lowilite 62, Songlight 6220, T 622LD, Tiangang BW 10LD, Tinuvin 622, Tinuvin 622LD, Tinuvin 622SF, Tinuvin LD 622, TNV 622, UV 622, UV 788, Uvinul 5062, Uvinul 5062H, XH 622LD |
| InChI formel | InChI=1S/C11H23NO2.C6H10O4/c1-10(2)7-9(14)8-11(3,4)12(10)5-6-13;1-9-5(7)3-4-6(8)10-2/h9,13-14H,5-8H2,1-4H3;3-4H2,1-2H3 |
| LEDER | CC(=O)CCC(=O)OCCN1C(CC(CC1(C)C)OC)(C)C |
| Molekylvikt (g/mol) | 347.45 |
| Formel vikt | 347.2308 |