missing translation for 'onlineSavingsMsg'
Läs mer

Poly(ethylene glycol) Monooleate (Technical Grade ~70%), TRC

Produktkod. 30412851
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412851 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30412851 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP6885055ML

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Polyoxyl Oleate
CAS 9004-96-0
Molekylformel (C2H4O)nC18H34O2
Kvantitet 5 mL
Synonym Adeka Estol OEG 204, Agnique PEG 400MO, Akyporox O 50, Alkamuls 400MO, Alkamuls A, Alkamuls A 2, Alkasurf O 14, Alkasurf O 75-9, AtlasG 2140, Blaunon EN 1507, CRL 1337, Cemulsol 1050, Chemax E 400MO, Chemester 300OC, ChimiponOCD, Cithrol 2MO, Cithrol PO, Crodet O 100, Dyapol G, Emalex 218, Emanon 4110, Emanon 4115, Emcol H 2A, Emcol H 31A, Emerest 2624, Emerest 2646, Emerest 2660, Empilan BP 100, Empilan BQ100, Emulan A, Emulphor 24, Estax 38 S.F, Ethofat O, Ethox MO 14, Ethoxylated oleic acid, Ethylan A 2, Ethylene oxide-oleic acid adduct, Extrex P 60, G 2142, G2143, G 2144, G 5507, H 2A, H 3549A, Hamadol OD 600, ITRI-PH 02, Interwet 33, Ionet MO 200, Ionet MO 400, Ionet MO 600, Ionet PO 600, KEOL15, Kessco PEG 1000MO, Kessco PEG 400MO, Konion MO 14, Koremul OA 9, Lannagol LF, Ligalub PEG 400MO, Lionon MO 60, Lipal 300W, Lipal30W, Lipal 400OL, Lipo-Peg 4O, MO 600, MYO 2, Macrogol oleate, Macrogol oleate 600, Mapeg 200, Marlosol OL 15, Marlosol OL 7, Mergital OA 6, Monooleate ester of polyethylene glycol, Newcol 170, Nikkol MYO 10, Nissan Nonion O 2, Noigen ES 120, 200, Nopalcol 1O, Oleic acid poly(oxyethylene) ester, Oleopal 1, Oleox 3, PEG 1000MO, PEG 200 oleate, PEG 200MO, PEG 300 monooleate, PEG-400 monoester with oleic acid, Pegnol O 14, Pegosperse 400MO, Peretex E 451D, Pionin D 2507A, Plasthall CPH 41N, Poly(ethylene glycol) oleic acidester, Poly(ethylene oxide) oleate, Poly(oxyethylene) monooleate, Poly(oxyethylene) oleate, Poly(oxyethylene)oleic acid ester, Polyethylene glycol monooleate, Polyethylene glycol oleate, Polyethylene oxide monooleate, Polyglycol monooleate, Polyglycol oleate, Polynon O 44, Polyoxyethylene ether oleate, Polyoxyethylene monooleic acid ester, Prodhyphore B, Radiasurf 7402, Rikemal OE 809, Rokacet O 7, S 1006, S 1132, Serdox NOG 264, Slovasol A, Standapol 2660, Surfonic E 400MO, Teric OF 6, Texafor M 6, Tilol 165, Trydet OA 7, Unichema A, Unisol 4-0, Wardol X, Witco 31, Witconol H 31A, Witconol H 35A, X539R
InChI formel InChI=1S/C20H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)23-19-18-21/h9-10,21H,2-8,11-19H2,1H3/b10-9-
LEDER O=C(OCCO)CCCCCCC/C=C/CCCCCCCC
Molekylvikt (g/mol) 326.51
Formel vikt 326.2821
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.