Läs mer
Poly[methylene(polyphenyl Isocyanate)] (Mn~340-400), TRC
Produktkod. 30003552
Change view
Kvantitet:
5 mL
10 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003552 10 mL
30005184 5 mL
30010139 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003552 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP68809510ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 9016-87-9
Molekylformel C15H10N2O2
Molekylvikt (g/mol) 250.25
Synonym Poly(methylenephenylene Isocyanate), Poly(phenylenemethylene Isocyanate)
LEDER O=C=NC1=CC=CC=C1.O=C=NC2=CC=CC=C2.O=C=NC3=CC=CC=C3.CCC.CCC
Specifikationer
Kemiskt namn eller material Poly[methylene(polyphenyl Isocyanate)]
Kvantitet 10 mL
InChI formel InChI=1S/C15H10N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h1-8H,9H2
Formel vikt 149.0477