missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(styrene sulfonic acid) sodium salt, M.W. 500,000, Thermo Scientific Chemicals
343.98 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Poly(styrene sulfonic acid) sodium salt is used in superplastifier in cement, ion exchange resins, membranes in fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to White |
Quantity | 25 g |
Solubility Information | Soluble in water and lower glycols. |
Physical Form | Powder |
Chemical Name or Material | Poly (styrene sulfonic acid) sodium salt, M.W. 500,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'