missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(styrene sulfonic acid) sodium salt, M.W. 70,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Poly(styrene sulfonic acid) sodium salt is used as Polyelectrolyte. It is also used as electroconductive and antistatic resin for electrographic and electrophotographic substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 50 g |
Solubility Information | Soluble in water,lower alcohols,glycols. |
Physical Form | Powder |
Chemical Name or Material | Poly(styrene sulfonic acid) sodium salt, M.W. 70,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'