Learn More
Kartell™ Plastilab™ Polyethylene Flat-Bottom Sample Vials
Catalog Number p-8005840
Click to view available options
Capacity (Metric):
1 mL
35 mL
Thickness (Metric):
0.98mm
1.6mm
General purpose containers for laboratory weighing and storage needs.
  • Convenient push-on attached hinged lids
  • Lightweight and unbreakable
  • Translucent
  • Suitable for storing food stuff
Closure Type Push on Hinged Lid
Product Type Flat Bottom Sample Vial
Capacity (Metric) 1 mL
Includes Push on hinged lid
Material Polyethylene
Thickness (Metric) 0.98mm