missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polyethylene rod, High Density, 19mm (0.75 in.) dia. 2.5cm

Brand:  Alfa Aesar™ 045180.BV

Additional Details : Weight : 0.01000kg

Quantity 30cm

 View more versions of this product

Product Code. 15466805

  • 259.36 SEK / 30cm


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

Polyethylene rod
High Density, 19mm (0.75 in.) dia.
MFCD00084423

Specifications

Polyethylene rod
Rod
Odorless
High Density, 19mm (0.75 in.) dia.
MFCD00084423
30cm
Videos
SDS