Learn More
Kartell™ Polyethylene Dropping Bottle Screw Caps
Click to view available options
Thread Size (Metric):
GL 18
GL 25
Length (Metric):
16mm
40mm
41mm
Description
Manufactured from polyethylene for strength and fit. Kartell™ Polyethylene Dropping Bottle Screw Caps, available in a variety of sizes and spout styles, provide a sturdy solution for laboratory dropping needs.
- Available in long or short dropping spout
- Fits standardized narrow and wide neck bottles
- Precision dropping for exact measurements with long spout
- Steady stream for filling small quantities with short spout
- Supplied with PE tip closure cap to avoid spillage
Specifications
Includes | PE cap for tip closure |
Material | Polyethylene |
Length (Metric) | 16mm |
Quantity | 100Pack |
Thread Size (Metric) | GL 18 |
For Use With (Equipment) | Narrow neck standardized bottles part. no. 15801 - 15821 - 15841 - 15861 - 15881 - 15901 and wide neck standardized bottles part. no. 1608 - 1610 |
Closure Type | Short Spout |