Learn More
Ark Plas™ Polyethylene Spiral Wrap
An ideal covering used to bundle and protect tubing and wires
Brand:  Ark Plas™ WXX77-TE1
Additional Details : Weight : 1.47600kg
 View more versions of this product
Product Code. 11808541
  • 1855.91 SEK / Each
In stock, ready to ship within 7 days: 1
Add to basket
Description
Description
  • Ark-Plas spiral wrap is manufactured from a lightweight, low cost LLDPE
  • Can be bundled around most items with a diameter of 1/16 in. to 7 in.
  • Specifications
    Specifications
    Black
    Polyethylene
    30 m
    Bundling Range: 5/8 to 4-1/2 in.
    Standard Spiral Wrap
    15.875 mm
    1.5748 mm