Learn More
BRAND™ Polyethylene Stopper
Polyethylene Stopper is suitable for use with centrifuge tubes.
139.00 SEK - 261.00 SEK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10099690
|
BRAND™
115368 |
26 mL Centrifuge Tubes (Art. No. 115346) |
139.00 SEK
Pack of 100 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
10314901
|
BRAND™
115372 |
75 mL Centrifuge Tubes (Art. No. 115352) |
261.00 SEK
Pack of 100 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
- Cap style stopper
- For centrifuge tubes
Specifications
Polyethylene | |
Stopper |