missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ark Plas Products™ Polyethylene Tubing
An economical choice for nonconductive tubing in the laboratory, providing good resistance to chemicals.
Brand: Ark Plas Products™ TEX83-TE0
Additional Details : Weight : 1.44000kg
Description
- Structure helps maintain flexibility and durability, even at low temperature
- Broad resistance to chemicals, can be used as a fluid line for liquids or gases
- Manufactured from resin, natural in color
- Extruded using state of the art inline process monitoring equipment
- Shipped bulk on cardboard spools
Specifications
Natural | |
Autoclavable | |
Polyethylene | |
300 m | |
1 Pack |
44 (Shore A) | |
9.52 mm | |
12.7 mm | |
1.57 mm |