Learn More
Kautex™ Polyethylene Wide Mouth Square Bottles
Manufactured with HDPE for strength and durability. Kautex™ Polyethylene Wide Mouth Square Bottles are ideal for packaging, storage and transporting media.
Brand:  Kautex™ 2000070539
Additional Details : Weight : 16.40000kg
 View more versions of this product
Product Code. 10168061
  • 21.95 SEK / Pack of 164
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
  • Easy filling, emptying and cleaning
  • Ribbed sides for a secure grip
  • Rectangular labelling field
  • Rectangular shape optimizes space utilisation
  • Leakproof
Specifications
Specifications
Wide Neck Bottle
500 mL
30.8 mm
HDPE
164/Cs.
145.5 mm
Natural
Packaging, Storage, Transport
164
Square