Läs mer
Polyhexamethyleneguanidine Hydrochloride, TRC
Produktkod. 30136839
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136839 5 g -
30135642 2.5 g 2.5g
18020730 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136839 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5702805G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57028-96-3
Molekylformel C9H21N3
Molekylvikt (g/mol) 171.28
Synonym 1,6-Hexanediamine-guanidine hydrochloride Copolymer, Akacid ForteGuanidine Hydrochloride-1,6-hexamethylenediamine Copolymer;Guanidine Hydrochloride-1,6-hexanediamine Copolymer;Guanidine Hydrochloride-hexamethylenediamine Copolymer;Hexamethylene Diamine-guanidine Hydrochloride Copolymer, Hygisoft, Metacide;Metacide (Fungicide), Agrosept Forte
IUPAC-namn 1,2-dibutylguanidine
LEDER Cl.NCCCCCCN.NC(=N)N
Specifikationer
Kemiskt namn eller material Polyhexamethyleneguanidine Hydrochloride
Analyt- eller komponentnamn Polyhexamethyleneguanidine Hydrochloride
Frakt skick Room Temperature
InChI formel InChI=1S/C9H21N3/c1-3-5-7-11-9(10)12-8-6-4-2/h3-8H2,1-2H3,(H3,10,11,12)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 5 g
Formel vikt 211.1564