Läs mer
Polyhyllin VI, TRC
Produktkod. 30422666
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30422666 100 mg
1 options
Produktkod. 30422666 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP689480100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Polyhyllin VI
CAS 55916-51-3
Molekylformel C39H62O13
Kvantitet 100 mg
Synonym (3β,25R)-17-hydroxyspirost-5-en-3-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-Glucopyranoside, 17-Hydroxyparis V, Paris VI, Paris Saponin VI, Pennogenin, 3-O-α-L-rhamnopyranosyl-(1→2)-β-D-glucopyranoside, Olygonatoside B3, Saponin Tb
InChI formel InChI=1S/C39H62O13/c1-18-8-13-38(47-17-18)20(3)39(46)27(52-38)15-25-23-7-6-21-14-22(9-11-36(21,4)24(23)10-12-37(25,39)5)49-35-33(31(44)29(42)26(16-40)50-35)51-34-32(45)30(43)28(41)19(2)48-34/h6,18-20,22-35,40-46H,7-17H2,1-5H3/t18-,19+,20-,22+,23-,24+,25+,26-,27+,28+,29-,30-,31+,32-,33-,34+,35-,36+,37+,38-,39-/m1/s1
LEDER O=CNc1ccc(C(O)=O)cc1
Molekylvikt (g/mol) 738.9