Läs mer
Polyinosinic Acid (90%), TRC
Produktkod. 30408810
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408810 10 g
30418398 5 g
30426994 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30408810 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP69897510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30918-54-8
Molekylformel C10H13N4O8P
Molekylvikt (g/mol) 348.21
Synonym NSC 120952, Poly(5'-inosinic Acid), Poly(IMP), Poly(rI), Polyinosinic acid, ((2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-oxo-3H-purin-9(6H)-yl)tetrahydrofuran-2-yl)methyl dihydrogen Phosphate
LEDER OP(OC)(OC[C@@H]1[C@@H](OC)[C@@H](O)[C@H](N2C(N=CNC3=O)=C3N=C2)O1)=O
Specifikationer
Kemiskt namn eller material Polyinosinic Acid
Kvantitet 10 g
InChI formel InChI=1S/C10H13N4O8P/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20)/t4-,6-,7-,10-/m1/s1
Formel vikt 348.0471