missing translation for 'onlineSavingsMsg'
Läs mer

Thermo Scientific Chemicals Polymixin B sulfate

Produktkod. 15425349
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1g
5g
Förpackningsstorlek:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15425349 5g -
15415349 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15425349 Leverantör Thermo Scientific Chemicals Leverantörsnummer J63074.06
 Begränsad artikel
I lager
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Polymixin B sulfate, CAS # 1405-20-5, is a short polypeptide with antibacterial activity against gram-negative bacteria. | CAS: 1405-20-5 | C48H84N16O17S | 1301.571 g/mol

Antibiotic for gram-negative bacteria Polymyxin B Sulfate is an antibiotic mixture active against gram (-) organisms. It is also reported to inhibit Ca2+ dependent PKC (protein kinase C).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Polymixin B sulfate is a mixture of B1 and B2 polypeptides obtained from different strains of Bacillus polymyxa, with antibacterial activity against gram-negative bacteria. It does not exhibit activity against fungi and gram-positive organisms
  • This compound inhibits Ca2+ dependent PKC (protein kinase C)
  • It can bind lipopolysaccharide (LPS) of the outer membrane of gram-negative bacteria and disrupts the cytoplasmic membrane by inducing large pores to allow nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane

Application

  • Polymixin B sulfate targets BAC-1.2F5 murine macrophages as an LPS (lipopolysaccharide) inhibitor
  • In in vitro research, this compound is used to prevent bacterial contamination of cell cultures
  • In microbiology research, it is used as a positive and negative control during microbicidal concentration (MCC) assay
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1405-20-5

Specifikationer

Innehåll och lagring Keep cold
Formatera Powder
Molekylvikt (g/mol) 1301.571
Produkttyp Polymixin B sulfate
Kvantitet 5g

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.