Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint enables smaller containers to be supported on top the existing reservior.
Brand: Poulten & Graf GmbH 101 09224
Additional Details : Weight : 0.50000kg
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of Polypropylene
- Adapter with ground joint
- Can be used for most chemicals and liquids
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers | |
White | |
Polypropylene | |
1 | |
Ground joint |
Autoclavable | |
Adapters for OPTIFIX and POLYFIX dispensers | |
OPTIFIX and POLYFIX dispensers | |
TS 24 |