Learn More
Wheaton™ Polypropylene Cap
Click to view available options
Color:
Red
White
Specifications
Closure Size | 20-400 mm |
Product Type | Cap |
Color | Red |
Material | Polypropylene |
Autoclavable | Autoclavable |