missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polypropylene Chromabond™ HR-XC SPE Column
Brand: Macherey-Nagel™ 730049
Additional Details : Weight : 0.20000kg
Specifications
1 mL | |
85 μm | |
No |
SPE | |
Solid Phase Extraction Column | |
65 to 75 Å |