Learn More
Greiner Bio-One Cryo.s Cryogenic Tubes with Internal Thread
Manufactured with high-grade polypropylene for strength and high thermal resistance. Greiner BioOne™ Cryo.s Cryogenic Tubes with Internal Threads withstand a temperature range of -196 to +121°C.
2175.00 SEK - 3040.00 SEK
Specifications
Product Type | Cryogenic Tube |
---|---|
Autoclavable | Not Autoclavable |
Material | Polypropylene |
Temperature Range (Metric) | -196°C to +121°C |
Sterility | Sterile |
Product Code | Brand | Closure Color | Bottom Shape | Capacity (Metric) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Bottom Shape | Capacity (Metric) | Includes | Price | Quantity & Availability | ||
10329910
|
Greiner Bio-One
121279 |
Blue | Round | 2 mL | White writing area; cap inserts for labeling |
3040.00 SEK
Pack of 500 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10563821
|
Greiner Bio-One
121278 |
Yellow | Round | 2 mL | White writing area; cap inserts for labeling |
3040.00 SEK
Pack of 500 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10450181
|
Greiner Bio-One
124274 |
Blue | Round | 5 mL | White writing area; cap inserts for labeling |
2175.00 SEK
Pack of 300 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Free of detectable human DNA, endotoxins, DNase and RNase
- Sterile, non-pyrogenic, non-cytotoxic, no heavy metals contained
- CE-marked (CE/IVD)
- As a safety precaution Cryo.s should be stored in a freezer or exclusively in the gas phase over the liquid nitrogen
- Suitable for shipment by air
- With white, scratch-resistant writing area
- 150 inserts per case (for additional labelling on the cap)
- Suitable for storage at -20°C, -80°C and in the vapour phase of LN2
- Multiple cap colours and working volumes
- Ultra-pure medical grade, USP class VI certified polypropylene w/o leachables
- Accessory storages boxes and working racks available
Specifications
Cryogenic Tube | |
Polypropylene | |
Sterile | |
Polypropylene |
Not Autoclavable | |
-196°C to +121°C | |
Yes |