Learn More
Eppendorf™ Polypropylene DNA LoBind Polypropylene Microcentrifuge Tube
Maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding with these tubes. Eppendorf™ Polypropylene DNA LoBind Tubes are ideal for sample preparation and long-term storage.
154.00 SEK - 275.00 SEK
Specifications
Material Polypropylene
Closure Material Polypropylene
Color Transparent
Max. Force 30000xG
For Use With (Application) DNA LoBind consumables are perfect for DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10686313 Eppendorf™
0030108035
0.5mL 275.00 SEK
Pack of 250 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 4
Add to basket
10051232 Eppendorf™
0030108051
1.5mL 154.00 SEK
Pack of 250 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 104
Add to basket
10031282 Eppendorf™
0030108078
2mL 187.00 SEK
Pack of 250 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 18
Add to basket
Description
Description
  • Free from DNA, DNase, RNase and ATP (PCR* clean)
  • Less than 1% loss of DNA /RNA at low concentrations and critical conditions (high salt, short DNA-fragments)
  • Special PP type manufactured under optimized conditions
  • Available as 0.5mL, 1.5mL and 2mL Safe-Lock™ tubes
  • High degree of transparency
Specifications
Specifications
Polypropylene
Transparent
DNA LoBind consumables are perfect for DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
Polypropylene
30000xG
250 Tubes (5 bags of 50)