Läs mer
Polypropylene Lift Pumps
Produktkod. 12937974
Encompass
Beskrivning:
Includes 1000 mm Downtube
Includes 900 mm Downtube
Förpackningsstorlek:
Varje
Produktkod. 12937974
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
For 20L/25L containers, capacity 0.33L per stroke. For 205L containers capacity 0.5L per stroke. Available in two sizes. With a 900mm flexible dispensing tube, except polypropylene 205L dispensing pump.
  • Manufactured with nylon for maximum resistance to acids and solvents
  • No metal part comes into contact with the liquid being pumped
  • PTFE seals fitted
  • Universal adapters fitted to provide secure seating in the necks of virtually all drums from 20L/25L to 205L capacities
  • Piston action ensures that the liquid flow is totally under control at all times during the action For dispensing solvents and acids in controlled amounts.
Specifikationer
Beskrivning Includes 1000 mm Downtube
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.