Läs mer
Pal International™ Deluxe Embossed Sole Fabric Overshoes
Produktkod. 12363996
Change view
Color: Blue
Produktkod.
12363996 Color: Blue
1 options
\r\n Utg\u00E5ngen artikel:\r\n
\r\n Den h\u00E4r artikeln utg\u00E5r ur v\u00E5rt sortiment n\u00E4r v\u00E5rt lager \u00E4r slut.\r\n
\r\n
Produktkod. 12363996 Leverantör Pal International™ Leverantörsnummer N68230
  • ONLINE EXCLUSIVE PRICE
    1080.00 SEK
    741.45 SEK / Paket med 200
    Save 338.55 SEK
    31% Av
I lager
\r\n Utg\u00E5ngen artikel:\r\n
\r\n Den h\u00E4r artikeln utg\u00E5r ur v\u00E5rt sortiment n\u00E4r v\u00E5rt lager \u00E4r slut.\r\n
\r\n
Designed for a better fit and improved comfort. Pal International™ Deluxe Embossed Sole Fabric Overshoes are especially suitable for labarotories or for visitors to hygienic manufacturing sites. Embossed PVC sole giving extra grip and durability.
    li>Manufactured from strong and durable non-woven polypropylene.
  • Higher spec overshoe designed for a better fit and improved comfort.
  • Especially suitable for more sensitive areas such as labarotories or for visitors to hygienic manufacturing sites.
  • Embossed sole gives extra grip and durability.
  • Elasticized band for easy wear and removal
  • Embossed sole provides extra grip and durability
  • DIsposable
  • One size fits all.
  • Package of 200
Specifikationer
Färg Blue
Klackstil Elasticated ankle
Material Polypropylene
Storlek Universal
Enda funktioner Embossed for grip and durability
Produkttyp Overshoe