missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Polypropylene Racks

Brand:  Tenak™ TE22361

Additional Details : Weight : 0.34000kg

 View more versions of this product

Product Code. 15270574

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Green
Rack
10 x 50 mL and 2 x 15 mL tubes
130 mm
Polypropylene
For 15 and 50mL Centrifuge Tubes
12 Pack
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers