Learn More
Brady™ Polypropylene Slippery Floor Warning Signs
Brand:  Brady™ 241882
Additional Details : Weight : 0.28600kg
Product Code. 11791995
  • 640.00 SEK / Each
Estimated Shipment: 14-05-2024
to see stock.
Specifications
Polypropylene
Warning sign for slippery floor
300 mm
Slippery Floor Warning Sign
1 Ea.
500 mm
Danger Slippery Floor Pictogram /Skid Risk