Learn More
Multiroir™ Polypropylene Modular Storage Block
Product Code. 11794581
Change view
Click to view available options
Capacity (Metric):
7.2 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
11794581 7.2 L Each
1 options
Product Code. 11794581 Supplier Multiroir™ Supplier No. 12109
  • ONLINE EXCLUSIVE PRICE
    1910.00 SEK
    920.84 SEK / Paket med 1
    Save 989.16 SEK
    51% Av
Beräknad leverans: 27-11-2026
Place the versatile Multiroir™ Polystyrene Modular Storage Block on a bench, attach it to the wall or suspend it under a shelf. Its total capacity comprised of various-sized drawers is 7.2 liters.
  • To be placed on bench, screwed to the wall or suspended under a shelf
  • Polystyrene drawer and ABS white walls
  • Delivered complete with transparent drawers
Specifications
Capacity (Metric) 7.2 L
Color Transparent
Material Polypropylene
Shape Rectangular
No. of Places 1 Each
Product Type Storage Block
Quantity 1 Each
Dimensions (L x W x H) 263 x 22 x 182 mm
Length (Metric) 263 mm
Height (Metric) Interior 182 mm