missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Polypropylene Tubing

Designed for indoor/outdoor applications requiring more rigid tubing. Ark Plas Products™ Polypropylene Tubing is extruded using state of the art in-line process monitoring equipment to ensure the highest quality.

Brand:  Ark Plas Products™ TFX84-TP0

Additional Details : Weight : 1.35900kg

 View more versions of this product

Product Code. 11753904

  • 1473.64 SEK / Each


In stock, ready to ship within 7 days: 2
Add to basket

You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.

Description

Description

Ark Plas tubing is available in a wide range of chemical and physical characteristics to fit most any application, dependably and economically.
  • Standard tubing is shipped bulk on cardboard spools
  • Manufactured in almost every increment from 1/16 to 3/8 in. I.D.
  • Metric sized tubing is also available from 2.5 to 6.5mm I.D.
  • Colors include Natural, Opaque Black, Blue, Red, Green, Yellow and White; and Transparent Blue, Red, Green and Yellow
Specifications

Specifications

Natural
Autoclavable
Polypropylene
300 m
1 Pack
80 (Shore A)
9.6 mm
12.7 mm
1.65 mm
Videos
SDS
Documents