Learn More
Macherey-Nagel™ Type N 9 Screw Neck Micro-Vials, PP with Inner Cone
Can be used on most HPLC and GC autosamplers. Macherey-Nagel™ Type N 9 Screw Neck Micro-Vials, PP with Inner Cone feature a polypropylene vial with a 0.3mL inner cone for micro-samples. They can be used with a range of screw closures with different septum materials/colors.
Brand: Macherey-Nagel™ 702009
This item is not returnable.
View return policy
Description
- Made of polypropylene, for pH-sensitive, aqueous samples
- Large range of screw closures (sold separately), including a magnetic version for the CTC GC PAL
- Bonded screw caps available (septa firmly connected, cannot be removed) so HPLC needles cannot push the septum into the vial
- Packaged as 100 pieces per PE bag
Specifications
Transparent | |
Polypropylene | |
Chromatography (GC, HPLC) | |
Screw Cap/Threads | |
Polyethylene Bag | |
32 mm | |
Autosampler, Screw Cap |
11.6 mm | |
0.3 mL | |
With Inner Cone and Screw Threads N 9 | |
100 | |
Screw Cap | |
100 Pk. |