Learn More
Kartell™ Plastilab™ Polypropylene Universal Test Tube Racks
Special design allows samples to be incubated at same temperature.
Brand:  Kartell™ 0056604
Additional Details : Weight : 2.78000kg
 View more versions of this product
Product Code. 12094448
  • 1875.00 SEK / Pack of 20
Estimated Shipment: 17-08-2021
Add to basket
Description
Description
Special design allows samples to be incubated at same temperature.
  • Autoclavable at 121°C
  • Large capacity
  • Can be used dry or wet in waterbaths
  • Does not rot, rust, or deform
  • Alphanumerical grid reference on top tier for easy identification
  • Color-coded for bath work
Specifications
Specifications
40
Autoclavable
246 mm
It can be used dry, wet, in water baths
Polypropylene
40
Yes, When Empty
Blue
20 mm
Tubes (20 mm)
1 Pack
Test Tube Rack